Formalin 10%, Q Path®, buffered
Pot, 40 ml in a 70 ml plastic pot, 50
Formaline klaar om fixatief te gebruiken voor weefsel stalen. Potjes en emmers kunnen gebruikt worden als opslag.
Formaldehyde 4% is ongeveer gelijk aan Formaline 10%.
: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
- :50-00-0
- :CH₂O
- :MFCD00003274
- :30.03 g/mol
- :1,013 g/cm³ (20 °C)
- :Kamertemperatuur
- :200-001-8
Documents
Cutsheet VWRQ10.07040WH0002.pdf Cutsheet VWRQ10.300150WH01.pdf Cutsheet VWRQFOR0070AF59001.pdf
Technical Drawing
FOR0070AF59001
Images 360
- Appearance:Colourless liquid
- Test formaldehyde:Passes test
- Density (20/4):1,012 - 1,014
- pH (20°C):6,8 - 7,2
- Verpakking:Pot
- VPE:50
- Info VPE:40 ml in a 70 ml plastic pot
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



