Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Bethanechol chloride
5 g
  ICNA0219016005
Bethanechol chloride
  ICNA0219016005
 :  590-63-6
Bethanechol chloride
5 g
 :  590-63-6
135,00 €
1 * 5 g
H2O: soluble 1.7 g/mL (stable for several days at 4 °C.); Ethanol: soluble 80 mg/mL (stable for several days at 4°C.) (Lit.)
Bethanechol Chloride is used to study beneficial effects of estrogen in a mouse model of cerebrovascular insufficiency.
 :  2-(Carbamoyloxy)-N,N,N-trimethyl-1-propanaminium Chloride
  •  :590-63-6
  •  :C₇H₁₇ClN₂O₂
  •  :MFCD00055224
  •  :196.675 g/mol
  • VPE:
    5 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.