Bethanechol chloride
5 g
ICNA0219016005
H2O: soluble 1.7 g/mL (stable for several days at 4 °C.); Ethanol: soluble 80 mg/mL (stable for several days at 4°C.) (Lit.)
Bethanechol Chloride is used to study beneficial effects of estrogen in a mouse model of cerebrovascular insufficiency.
: 2-(Carbamoyloxy)-N,N,N-trimethyl-1-propanaminium Chloride
- :590-63-6
- :C₇H₁₇ClN₂O₂
- :MFCD00055224
- :196.675 g/mol
- VPE:5 g