Orders LinkContactUsLinkComponent
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
roos
Catalogus # FTEC110-04-10LMI
Leverancier:   FA TECH DIAGNOSTICS
Embedding cassettes with disposable lid
Catalogus # FTEC110-04-10LMI
Leverancier:  FA TECH DIAGNOSTICS
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
roos
Price
101,00 €
Log in voor beschikbaarheid
1 * 1.000 ST
Product Details & Documents
Embedded cassettes with hinged lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
Ordering information: Available in different colours.
Packaging: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Specificaties
  • Kleur:
    roos
  • Seq:
    70