Cuvettes and holders
Four-position cell changer, 10 - 50 mm
Catalogus # SCLI80-2106-01
Leverancier: BIOCHROM
Ga naar:
Product Details & Documents
Cuvettes for spectrophotometers designed to hold samples during spectroscopic experiments.
Different pathlengths and formats are available.
Specificaties
- Omschrijving:Four-position cell changer, 10 - 50 mm
- Seq:06