Cuvettes and holders
Cell holder for 10 - 50 mm pathlength cuvettes
Catalogus # SCLI80-2106-07
Leverancier: BIOCHROM
Ga naar:
Product Details & Documents
Cuvettes for spectrophotometers designed to hold samples during spectroscopic experiments.
Different pathlengths and formats are available.
Specificaties
- Omschrijving:Cell holder for 10 - 50 mm pathlength cuvettes
- Seq:05