Cuvettes and holders
Semi micro glass cell, 800 µl, 10 mm
Catalogus # SCLI80-2004-15
Leverancier: BIOCHROM
Ga naar:
Product Details & Documents
Cuvettes for spectrophotometers designed to hold samples during spectroscopic experiments.
Different pathlengths and formats are available.
Specificaties
- Lichtweg:10 mm
- Volume:800 µl
- Omschrijving:Semi micro glass cell
- Seq:03