Orders LinkContactUsLinkComponent
Cables
InLab® cable S7-DIN, 1,2 m
Catalog # 661-0261
Supplier:   Mettler - Toledo
Cables
Catalog # 661-0261
Supplier:  Mettler - Toledo
Cables
Cables
InLab® cable S7-DIN, 1,2 m
Price
70,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Specifications
  • Omschrijving:
    InLab® cable S7-DIN
  • Lengte:
    1,2 m
  • Seq:
    162