Microscope slides for printers
blauw
Catalogus # KLINKP-SIL-3057
Leverancier: KLINIPATH
Ga naar:
Product Details & Documents
Microscope slides for slide printers.
- Frosted ends
- 90° ground edges
- Finished clipped corners of 1 mm
Allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
Specificaties
- Kleur:blauw
- Seq:09