PARALLEL GUIDE 100MM 1 * 1 ST
KPXX38073
PARALLEL GUIDE 100MM 1 * 1 ST
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Natriumhydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals
Bufferoplossingen, gebruiksklaar, waarnaar wordt verwezen bij 20 °C, CertiPUR®, Supelco®
#: 1.09439.1000
: Merck



