Embedding cassettes, without lid
lichtblauw
Catalogus # KLIN310-08S
Leverancier: KLINIPATH
Ga naar:
Product Details & Documents
Every cassette maintain its stability, rigidness and shape during processing of tissues.
: Hoppers contain 75 standard cassettes and are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes and are specially packed for Sakura® / Leica® cassette printers.
Specificaties
- Kleur:lichtblauw
- Seq:08