Slide label rolls, polyester
Slide label, Polyester, wit, 24×19 mm, On roll
KLIN1513-K
These slide labels are made of polyester and are chemical resistant.
- Kleur:wit
- Omschrijving:Slide label
- Afmetingen:24×19 mm
- Materiaal:Polyester
- Seq:005
- Verpakt:On roll