Embedding cassettes with disposable lid
wit
Catalogus # FTEC110-01-10LM
Leverancier: FA TECH DIAGNOSTICS
Embedding cassettes with disposable lid
Catalogus # FTEC110-01-10LM
Leverancier: FA TECH DIAGNOSTICS
Ga naar:
Product Details & Documents
Embedded cassettes with hinged lid.
- Cassettes maintain their stability, rigidness and shape during processing of tissues
: Available in different colours.
: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Specificaties
- Kleur:wit
- Seq:39