Orders LinkContactUsLinkComponent
Objectives for CKX53
Dry objective with pre-centre phase contrast slider, Ph 20×/0,4 (dry), 3,2 mm
Catalogus # OLYMN5229300
Leverancier:   OLYMPUS OPTICAL
Objectives for CKX53
Catalogus # OLYMN5229300
Leverancier:  OLYMPUS OPTICAL
Objectives for CKX53
Objectives for CKX53
Dry objective with pre-centre phase contrast slider, Ph 20×/0,4 (dry), 3,2 mm
Price
740,36 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Vergroting:
    Ph 20×/0,4 (dry)
  • Omschrijving:
    Dry objective with pre-centre phase contrast slider
  • Seq:
    022
  • Werkafstand:
    3,2 mm