Order Entry
Belgium
Orders LinkContactUsLinkComponent
Formaldehyde 4%

Formaldehyde 4%

10 L
Catalogus # VWRK4078.9010
Irriterend
Gezondheid
Leverancier:   KLINIPATH
Formaldehyde 4%
Catalogus # VWRK4078.9010
Leverancier:  KLINIPATH
CAS-nummer:  50-00-0
Formaldehyde 4%

10 L

CAS-nummer:  50-00-0
Price
39,40 €
Log in voor beschikbaarheid
1 * 10 L
Product Details & Documents

Formaldehyde 4% is approximately equal to Formalin 10%

  • pH 7
  • Phosphate buffered
Synoniemen:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS-nummer:50-00-0
  • Formule:CH₂O
  • MDL-nummer:MFCD00003274
  • Molecular Weight:30.03 g/mol
Specificaties

Specificatie testresultaten

  • Formaldehyde (HCHO) g/l:
    38 - 42
  • pH at 25 °C:
    6.8 - 7.2
  • Sodium (Na):
    61 - 71 mmol/l

Specificaties

  • VPE:
    10 L

Specificaties