Embedding cassettes with disposable lid
oranje
Catalogus # KLIN110-06T
Leverancier: KLINIPATH
Ga naar:
Product Details & Documents
Embedded cassettes with hinged lid.
- Cassettes maintain their stability, rigidness and shape during processing of tissues
Ordering information: Available in different colours.
Packaging: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Specificaties
- Kleur:oranje
- Seq:30
- Cat. No.:KLIN110-06T