Frocks, PP
groen, XXL
Catalogus # HOPE40.301V/05
Leverancier: Medicom
Ga naar:
Product Details & Documents
These single-use hygiene frocks made from PP protect the working environment from particles coming from the operator's body (dander, hair, perspiration, etc.).
- With collar and long sleeves
- Elasticated cuffs
- Snap buttons
- Without pockets
Weight: 40 g/m²
Documents
Specificaties
- Confectiemaat:XXL
- Kleur:groen
- Cat. No.:HOPE40.301V/05
- Kledingstuk toepassing:General Purpose|Industrial
- Sluitingstype:Met drukknopen
- Zakken:zonder zakken
- Gewicht:40 g/m²