Orders LinkContactUsLinkComponent
BioSample cards
BioSample cards
BioSample cards with cover, 0,07 ml
Catalogus # BINZ8.460.0001.B-N
Leverancier:   Ahlstrom
BioSample cards
Catalogus # BINZ8.460.0001.B-N
Leverancier:  Ahlstrom
BioSample cards
BioSample cards
BioSample cards with cover, 0,07 ml
Price
168,00 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
These BioSample cards are designed for microvolume sampling, transport and ambient storage of biological samples. These cards are made of white and pure absorbent fibers and contain no wet-strength additives or chemicals. This feature ensures reproducible collection and minimise possible interferences for reliable analysis of targeted analytes like small molecules, proteins, nucleic acids or vitamins.
  • Reliable and reproducible sample collection
  • Even and uniform sample distribution
  • Customisable design and printing
  • Rigorous quality control
  • Each card has five/eight spots
General applications include protein-based analysis, lipid-based analysis, genomics, human identification and forensics. Indicated cards can also be used for HIV viral load testing and infectious disease screening (including Hepatitis B, C and Syphilis).
Certificeringen: Manufacturing according to ISO 13485:2016 and ISO 9001:2015 standards.
In Vitro Diagnostic medical device in compliance with the Directive 98/79/EC.
Bestelinformatie: Drying racks are available to facilitate drying of the cards at ambient temperature.
Let op: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Specificaties
  • Volume:
    0,07 ml
  • Omschrijving:
    BioSample cards with cover
  • Seq:
    01
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.