Microplates, opaque OptiPlates™
OptiiPlate™-384 microplates, 384, wit, Case of 50
PERK6007290
Marketsource
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
- Made of polystyrene
- Aant. wells:384
- Verpakt:Case of 50
- Kleur:wit
- Omschrijving:OptiiPlate™-384 microplates
- Seq:04