Orders LinkContactUsLinkComponent
O-rings
O-ring, DN 100, Siliconen
  NEUB5190-100
O-rings
  NEUB5190-100
O-rings
O-ring, DN 100, Siliconen
39,90 €
1 * 1 ST
Silicone O-rings suitable for flat flanges.
  • Omschrijving:
    O-ring, DN 100
  • Materiaal:
    Siliconen
  • Seq:
    02