Accessories for QUANTOFIX®
URYXXON® Controlestrip; brutogewicht 67 g, verpakkingsafmetingen 161 × 63 × 30 mm
Leverancier: MACHEREY-NAGEL
Ga naar:
Product Details & Documents
The accessories for the QUANTOFIX® product line help to improve the experience while using the test strips.
Useful accessories for QUANTOFIX® products
Specificaties
- Omschrijving:URYXXON® Controlestrip; brutogewicht 67 g, verpakkingsafmetingen 161 × 63 × 30 mm
- Seq:04
- Cat. No.:MANA930988