Orders LinkContactUsLinkComponent
Zanamivir, Reference Standards
Zanamivir, Reference Standards
Zanamivir Resolution Mixture, USP reference standard, 0.25 mg, Vial, 250 µG
Catalogus # 1724066.
Leverancier:   USP
Zanamivir, Reference Standards
Catalogus # 1724066.
Leverancier:  USP
CAS-nummer:  
Zanamivir, Reference Standards
Zanamivir Resolution Mixture, USP reference standard, 0.25 mg, Vial, 250 µG
CAS-nummer:  
Price
609,00 €
Log in voor beschikbaarheid
1 * 250 µG
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Zanamivir Resolution Mixture: Mixture of zanamivir and talo-zanamivir
Certificeringen: US Pharmacopea Reference Standard
Specificaties
Specificaties
  • Verpakking:
    Vial
  • VPE:
    250 µG
  • Standaard:
    USP reference standard
  • Omschrijving:
    Zanamivir Resolution Mixture
  • Info VPE:
    0.25 mg
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.