AdvanTip™ Pipette Tips
Standard, 2 - 300 µl, -, 250, 300 μl pipettes, Racked, 96 tips/rack
Catalogus # HAMI68009-11
Leverancier: HAMILTON BONADUZ
Ga naar:
Product Details & Documents
AdvanTip™ precision pipette tips are perfect for routine, high-throughput work where an extensive range of universal tips and multiple packaging options are needed to meet laboratory requirements.
- Made from chemically resistant, virgin PP for excellent clarity and smooth surfaces
- Certified free from RNase, DNase, and pyrogens
- Extended length (only some tips): Prevent sample cross-contamination and protect pipettes from contamination when pipetting into long, narrow tubes or deep well plates
- Low retention tips (only some tips): Feature walls that maximise sample recovery; ideal for quantitative PCR, valuable samples and reagents
- Universal fit, can be used with Hamilton SoftGrip™ manual pipettes
Specificaties
- Verpakt:Racked, 96 tips/rack
- Steriel:-
- Volume:2 - 300 µl
- Seq:02
- Tip stijl:Standard
- Voor gebruik met:250, 300 μl pipettes
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.