Mobile storage cabinets
Mobile storage cabinet, with 2 drawer of 300 mm height, lockable, 2 drawers, 450×516×740 mm
Catalogus # KOTT315-00145
Leverancier: KOETTERMANN
Ga naar:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specificaties
- Omschrijving:Mobile storage cabinet, with 2 drawer of 300 mm height, lockable
- Seq:20
- b×d×h:450×516×740 mm
- Deuren:2 drawers
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



