Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Mobile storage cabinets

Mobile storage cabinets

Mobile storage cabinet, with 3 drawer of 150 mm height, pencil tray, 3 drawers, 450×516×590 mm
Catalog # KOTT315-00144
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00144
Supplier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets

Mobile storage cabinet, with 3 drawer of 150 mm height, pencil tray, 3 drawers, 450×516×590 mm

Price
1 170,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Epoxy powder coated steel cabinets

  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Omschrijving:
    Mobile storage cabinet, with 3 drawer of 150 mm height, pencil tray
  • Seq:
    09
  • b×d×h:
    450×516×590 mm
  • Deuren:
    3 drawers
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.