Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, lockable, 2 door, 900×516×740 mm
  KOTT315-00139
Mobile storage cabinets
  KOTT315-00139
Mobile storage cabinets
Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, lockable, 2 door, 900×516×740 mm

 
1 050,00 €
   
1 * 1 ST
 

Epoxy powder coated steel cabinets

  • Optional locks
  • Fitted with castors
  • Removable shelves
 
  • Omschrijving:
    Mobile storage cabinet, with 1 drawer of 150 mm height, lockable
  • Seq:
    33
  • b×d×h:
    900×516×740 mm
  • Deuren:
    2 door