Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, lockable, 2 door, 900×516×740 mm
Catalogus # KOTT315-00137
Leverancier:   KOETTERMANN
Mobile storage cabinets
Catalogus # KOTT315-00137
Leverancier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, lockable, 2 door, 900×516×740 mm
Price
819,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specificaties
  • Omschrijving:
    Mobile storage cabinet, lockable
  • Seq:
    31
  • b×d×h:
    900×516×740 mm
  • Deuren:
    2 door
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.