Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, 2 door, 900×516×740 mm
Catalogus # KOTT315-00136
Leverancier:   KOETTERMANN
Mobile storage cabinets
Catalogus # KOTT315-00136
Leverancier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, 2 door, 900×516×740 mm
Price
762,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specificaties
  • Omschrijving:
    Mobile storage cabinet
  • Seq:
    30
  • b×d×h:
    900×516×740 mm
  • Deuren:
    2 door