Mobile storage cabinets
Mobile storage cabinet, With front open, 900×500×740 mm
Catalogus # KOTT315-00133
Leverancier: KOETTERMANN
Ga naar:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specificaties
- Omschrijving:Mobile storage cabinet
- Seq:27
- b×d×h:900×500×740 mm
- Deuren:With front open