Mobile storage cabinets
Mobile storage cabinet, with 1 drawer of 150 mm height, lockable, 1 door right, 450×516×740 mm
Catalog # KOTT315-00125
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Omschrijving:Mobile storage cabinet, with 1 drawer of 150 mm height, lockable
- Seq:17
- b×d×h:450×516×740 mm
- Deuren:1 door right