Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet with 1 shelf, With front open, 450×500×590 mm
Catalog # KOTT315-00107
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00107
Supplier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet with 1 shelf, With front open, 450×500×590 mm
Price
347,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Omschrijving:
    Mobile storage cabinet with 1 shelf
  • Seq:
    01
  • b×d×h:
    450×500×590 mm
  • Deuren:
    With front open
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.