Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinet, with 1 drawer of 150 mm height, lockable, 2 doors, 1200×516×740 mm
Catalogus # KOTT315-00103
Leverancier:   KOETTERMANN
Mobile storage cabinets
Catalogus # KOTT315-00103
Leverancier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, with 1 drawer of 150 mm height, lockable, 2 doors, 1200×516×740 mm
Price
1 160,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specificaties
  • Omschrijving:
    Mobile storage cabinet, with 1 drawer of 150 mm height, lockable
  • Seq:
    48
  • b×d×h:
    1200×516×740 mm
  • Deuren:
    2 doors