Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, 2 doors, 1200×516×590 mm
Catalog # KOTT315-00095
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00095
Supplier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, 2 doors, 1200×516×590 mm
Price
736,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Omschrijving:
    Mobile storage cabinet
  • Seq:
    40
  • b×d×h:
    1200×516×590 mm
  • Deuren:
    2 doors