NMR sample tube washer
NMR sample tube washer, acc. to Swiegot, complete, to fit laboratory bottles with GL 45 thread
Catalogus # WITG2.900.032
Leverancier: witeg Labortechnik
Ga naar:
Product Details & Documents
Complete with 2 PTFE-adapters to fir 5 cuvets or NMR tubes of 5 and 10 mm diameter in different lengths. Vacuum operated washers allowing the individual washing of 1 to 5 tube, complete with caps and FEPcappilaries (box 1 pce.).
Specificaties
- Omschrijving:NMR sample tube washer, acc. to Swiegot, complete, to fit laboratory bottles with GL 45 thread
- Seq:02
- Cat. No.:WITG2.900.032