Orders LinkContactUsLinkComponent
NMR sample tube washer
NMR sample tube washer
NMR sample tube washer, acc. to Swiegot, complete, to fit bottles with socket ST 29/32
Catalog # WITG2.900.029
Supplier:   witeg Labortechnik
NMR sample tube washer
Catalog # WITG2.900.029
Supplier:  witeg Labortechnik
NMR sample tube washer
NMR sample tube washer
NMR sample tube washer, acc. to Swiegot, complete, to fit bottles with socket ST 29/32
Price
232,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Complete with 2 PTFE-adapters to fir 5 cuvets or NMR tubes of 5 and 10 mm diameter in different lengths. Vacuum operated washers allowing the individual washing of 1 to 5 tube, complete with caps and FEPcappilaries (box 1 pce.).
Specifications
  • Omschrijving:
    NMR sample tube washer, acc. to Swiegot, complete, to fit bottles with socket ST 29/32
  • Seq:
    01