Basic fuchsin 70+%, pure, certified
Glass bottle, 500 g
Leverancier: Thermo Fisher Scientific
Basic fuchsin 70+%, pure, certified
Catalogus # ACRO401695000
Leverancier: Thermo Fisher Scientific
CAS-nummer: 632-99-5
Ga naar:
Product Details & Documents
Synoniemen: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
- CAS-nummer:632-99-5
- Formule:C₂₀H₂₀ClN₃
- MDL-nummer:MFCD00012569
- Molecular Weight:337.85 g/mol
- Merck index:12,05686
- Smelten Pt:250 °C (ontbinding)
- Dichtheid:1,22 g/cm³ (20 °C)
- Flits Pt:200 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:211-189-6
Documents
ACRO40169
SDS
Specificaties
Specificatie testresultaten
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥70.0 %
- UV Lambda max:547 to 552 nm (in 50% ethanol)
- Absorption Ratio P-15/P+15:1.16 to 1.35
- BSC Certification:Passes
Specificaties
- Verpakking:Glass bottle
- VPE:500 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



