Basic fuchsin 70+%, pure, certified
Glass bottle, 100 g
Leverancier: Thermo Fisher Scientific
Basic fuchsin 70+%, pure, certified
Catalogus # ACRO401691000
Leverancier: Thermo Fisher Scientific
CAS-nummer: 632-99-5
Ga naar:
Product Details & Documents
Synoniemen: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
- CAS-nummer:632-99-5
- Formule:C₂₀H₂₀ClN₃
- MDL-nummer:MFCD00012569
- Molecular Weight:337.85 g/mol
- Merck index:12,05686
- Smelten Pt:250 °C (ontbinding)
- Dichtheid:1,22 g/cm³ (20 °C)
- Flits Pt:200 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:211-189-6
Documents
ACRO40169
SDS
Specificaties
Specificatie testresultaten
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥70.0 %
- UV Lambda max:547 to 552 nm (in 50% ethanol)
- Absorption Ratio P-15/P+15:1.16 to 1.35
- BSC Certification:Passes
Specificaties
- Verpakking:Glass bottle
- VPE:100 g



