L(-)-Mannose 99+%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
- CAS Number:10030-80-5
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00136021
- Molecular Weight:180.16 g/mol
- Boiling Pt:376 °C (1013 hPa)
- Melting Pt:129…131 °C
- Storage Temperature:Kamertemperatuur
- EINECS:233-080-2
Documents
ACRO22759
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- HPLC:≥99.0 % (ELSD)
- Specific optical rotation:-13.5° to -15.5° (20°C, 589 nm) (c=4, water, NH3)
- Heavy metals (as Pb):≤10 ppm
Specifications
- Verpakking:Glass bottle
- VPE:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



