D-(-)-N-Methylglucamine 99%
Plastic drum, 2,5 kg
Leverancier: Thermo Fisher Scientific
D-(-)-N-Methylglucamine 99%
Catalogus # ACRO126840025
Leverancier: Thermo Fisher Scientific
CAS-nummer: 6284-40-8
Ga naar:
Product Details & Documents
Synoniemen: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
- CAS-nummer:6284-40-8
- Formule:H₃CNHCH₂(CH(OH))₄CH₂OH
- MDL-nummer:MFCD00004707
- Molecular Weight:195.22 g/mol
- Merck index:12,06154
- Smelten Pt:128…132 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:228-506-9
Documents
Specificaties
Specificatie testresultaten
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:128 to 132 °C
- Loss on drying:≤1 % (1 g, 100 °C, 3 hrs)
- Sulfated ash:≤0.1 %
- UV:(20 % w/v in water)
- Specific optical rotation:-15.7° to -17.3° (20 °C, 589 nm) (c=10, H2O) (on dried substance)
- Titration with HClO4:≥98.5 % (On dried substance)
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤150 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
Specificaties
- Verpakking:Plastic drum
- VPE:2,5 kg
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



