Maleïnezuur 99%
Plastic drum, 5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:110-16-7
- Formula:C₄H₄O₄
- MDL Number:MFCD00063177
- Molecular Weight:116.07 g/mol
- Merck Index:13,05726
- Boiling Pt:256 °C (1013 hPa)
- Melting Pt:134…138 °C
- Density:1.59 g/cm³ (20 °C)
- Flash Pt:127 °C
- Storage Temperature:Kamertemperatuur
- EINECS:203-742-5
- UN:3261
- ADR:8,III
Documents
ACRO12523
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder or granules and/or chunks
- Infrared spectrum:Conforms
- Melting point:130 to 138 °C
- Water:≤2 %
- HPLC:≥98.5 %
- Titration with NaOH:98.5 to 101.5 % (on dry substance)
- Heavy metals (as Pb):≤10 ppm
Specifications
- Verpakking:Plastic drum
- VPE:5 kg



