3,5-Dimethoxybenzaldehyde 98%
Glass bottle, 50 g
- :7311-34-4
- :C₉H₁₀O₃
- :MFCD00003366
- :166.18 g/mol
- :151 °C (16 mmHg)
- :44…48 °C
- :>112 °C
- :Kamertemperatuur
- :230-772-6
- Appearance:White to beige Crystalline solid
- Infrared spectrum:Conforms
- Melting point:44 to 48 °C
- GC:≥97.5 %
- Verpakking:Glass bottle
- VPE:50 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



