Isonicotinic acid 99%
Plastic bottle, 2,5 kg
Leverancier: Thermo Fisher Scientific
Isonicotinic acid 99%
Catalogus # ACRO122590025
Leverancier: Thermo Fisher Scientific
CAS-nummer: 55-22-1
Ga naar:
Product Details & Documents
Synoniemen: Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
- CAS-nummer:55-22-1
- Formule:C₆H₅NO₂
- MDL-nummer:MFCD00006429
- Molecular Weight:123.11 g/mol
- Merck index:13,05206
- Kokend Pt:260 °C (15 mmHg)
- Smelten Pt:310…315 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:200-228-2
Specificaties
Specificatie testresultaten
- Appearance:White to cream Powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥98.5 %
- HPLC:≥97.5 %
- Specific optical rotation:-190° to -204° (20°C, 589 nm) (c=2,acetone)
Specificaties
- Verpakking:Plastic bottle
- VPE:2,5 kg