D-(+)-Malic acid 98%
Glass bottle, 25 g
: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
- :636-61-3
- :C₄H₆O₅
- :MFCD00004245
- :134.09 g/mol
- :13,05307
- :322 °C (1013 hPa)
- :99…104 °C
- :Kamertemperatuur
- :211-262-2
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:99 to 104 °C
- Water:≤1 % (K.F.)
- Sulfated ash:≤0.1 %
- HPLC:≤1 % Fumaric acid
- Specific optical rotation:+1.8° to +2.6° (20 °C, 589 nm) (c=3, Water)
- Titration with NaOH:≥97.5 %
- Verpakking:Glass bottle
- VPE:25 g



