Phthalaldehyde 98+%
Glass bottle, 500 g
Leverancier: Thermo Fisher Scientific
Phthalaldehyde 98+%
Catalogus # ACRO131085000
Leverancier: Thermo Fisher Scientific
CAS-nummer: 643-79-8
Ga naar:
Product Details & Documents
Synoniemen: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
- CAS-nummer:643-79-8
- Formule:C₈H₆O₂
- MDL-nummer:MFCD00003335
- Molecular Weight:134.13 g/mol
- Kokend Pt:83…84 °C (1hPa)
- Smelten Pt:54…57 °C
- Dichtheid:1,13 g/cm³ (20 °C)
- Flits Pt:132 °C
- Temperatuur bij opslag:Koelkast
- EINECS:211-402-2
- UN:2923
- ADR:8,II
Documents
Specificaties
Specificatie testresultaten
- Appearance:Yellow Powder or crystals and/or lumps
- Infrared spectrum:Conforms
- Melting point:54 to 58 °C
- GC:≥98.0 %
Specificaties
- Verpakking:Glass bottle
- VPE:500 g