Samplers, U-D Master
Single style sampler, 300 mm, 5, 25 mm
Catalogus # SAMP1230A-300
Leverancier: Sampling Systems
Ga naar:
Product Details & Documents
U-D Master is designed for sampling free flowing powders. By simply changing the sample insert cells, different volumes can be sampled.
- Operation of the unit dose powder sampler is very easy
- Push the sampler: At the required depth rotate the handle, which will take the sample
- After removing the sampler the sample inserts can be removed for easy emptying of the sample
Single version takes one sample from each location.
Double version takes two samples from each location.
Triple version takes three samples from each location.
Specificaties
- Ø:25 mm
- Omschrijving:Single style sampler
- Lengte:300 mm
- Seq:01
- Segmenten:5
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 0,2 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 0,9 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,0 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,1 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,3 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,2 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,4 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,5 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,6 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,7 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,8 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 1,9 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,0 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,2 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,3 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,4 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,5 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,6 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,7 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,9 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,0 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,2 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,3 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,1 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,4 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,5 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,6 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,8 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,9 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 4,0 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 3,7 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 0,3 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 0,4 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 0,5 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 0,7 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 0,6 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,8 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, blank
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 0,1 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 0,8 mlSample insert, stainless steel, 0,8 mlSAMP1230T-08Log in voor beschikbaarheid1 * 1 Pce - 136,00 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Sample insert, stainless steel, 2,1 ml