Mop frame, aluminium
Mop frame, aluminium, ClinoConnect lock, 400 mm
Catalogus # PPSA2009161
Leverancier: PFENNIG REINIGUNGSTECHNIK
Ga naar:
Product Details & Documents
Aluminium mop frame with reduced weight.
- Mechanics made of stainless steel, plate made of aluminium
- Autoclavable
- Contactless mop drop with MopDrop
Specificaties
- Omschrijving:Mop frame, aluminium, ClinoConnect lock
- Lengte:400 mm
- Seq:03