Toebehoren
Afdichtmat voor deepwell-plaat 96/2000 µl
Catalogus # EPPE0030127.960
Leverancier: EPPENDORF
Ga naar:
Product Details & Documents
Secure your samples with confidence using Eppendorf® Sealing mats, the ideal solution for incubation and short-term storage needs. These reusable mats can be autoclaved and come in a 'PCR Clean' purity grade (free of RNase, DNase, Human DNA and PCR inhibitors).
- Sealing mats are reusables and can be autoclaved
- Suitable for short-term storage
Documents
Specificaties
- Omschrijving:Afdichtmat voor deepwell-plaat 96/2000 µl
- Seq:02