Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Toebehoren
Toebehoren
Afdichtmat voor deepwell-plaat 96/2000 µl
Catalogus # EPPE0030127.960
Leverancier:   EPPENDORF
Toebehoren
Catalogus # EPPE0030127.960
Leverancier:  EPPENDORF
Toebehoren
Toebehoren
Afdichtmat voor deepwell-plaat 96/2000 µl
Price
180,00 €
Log in voor beschikbaarheid
1 * 50 ST
Product Details & Documents
Secure your samples with confidence using Eppendorf® Sealing mats, the ideal solution for incubation and short-term storage needs. These reusable mats can be autoclaved and come in a 'PCR Clean' purity grade (free of RNase, DNase, Human DNA and PCR inhibitors).
  • Sealing mats are reusables and can be autoclaved
  • Suitable for short-term storage
Specificaties
  • Omschrijving:
    Afdichtmat voor deepwell-plaat 96/2000 µl
  • Seq:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.