Solid phase extraction disks, Resprep®
Solid phase extraction disks, Resprep® C18, 90 mm
Catalogus # REST25988
Leverancier: Restek
Ga naar:
Product Details & Documents
These glass fiber disks are embedded with C18 or C8 bonded silica. Deep-pore design reduces clogging and allows faster flow rates. Semivolatile organic compounds can be etracted.
Certifications: Meet the requirements for EPA methods 525.1, 506, 550.1 and 549.1.
Specificaties
- Ø:90 mm
- Omschrijving:Solid phase extraction disks, Resprep® C18
- Seq:03