Inoculating loops, PS, calibrated
Inoculating loop, PS, sterile, 10 µl, blauw
Supplier: DELTALAB
Jump to:
Product Details & Documents
These calibrated, dual use inoculating loops offer a loop at one end to facilitate microbiological streaking, and a fine tip at the other specially designed for counting colonies.
- High robustness and flexible
- Colour-coded for ease of size identification
- Eliminates the need for flaming
- Sterilised by radiation
Documents
Specifications
- Capaciteit:10 µl
- Kleur:blauw
- Omschrijving:Inoculating loop, PS, sterile