Safety Spectacles, Silium+
Anti-UV (2C-1,2), anti-fog, anti-scratch, antraciet, Clear PC
Catalogus # 111-1625
Leverancier: Bollé Safety
Ga naar:
Product Details & Documents
Inspired from SILIUM, SILIUM + are comfortable and ultra lightweight PC safety glasses. SILIUM + is an excellent choice, combining style, ergonomics and performances for extended use.
- Adjustable bridge
- Non-slip bridge
- Straight, highly flexible ergonomic temples
- PLATINUM coating
Weight: 21 g
Certifications: EN 166 FT1, EN 170, EN 172 (smoke lens).
Documents
BOLESILPPSI_TDS_EN
Datasheet
Specificaties
- Type:Anti-UV (2C-1,2), anti-fog, anti-scratch
- Kleur:antraciet
- Beglazing:Clear PC