Racks for test tubes
20 mm, 40 (4×10), PE, 246×107×64 mm
Catalogus # HACH2497902
Leverancier: Hach
Ga naar:
Product Details & Documents
These test tube racks features a special blend of polyethylene which sinks in water, assuring non-floating racks when immersed.
- Stackable
- Good chemical resistance
- Good temperature resistance
Specificaties
- Boring Ø:20 mm
- Aant. gaten:40 (4×10)
- Materiaal:PE
- Seq:01
- b×d×h:246×107×64 mm